Learn More
Meso Scale Discovery SPLEX HUM IL2 KitQP 25 PL
SDP
Catalog No. 5000081807
Encompass
The Meso Scale Discovery S-PLEX Human IL-2 Kit is a multiplex immunoassay that measures human interleukin-2 (IL-2) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay takes 3 5 hours to complete and has a detection range of 0 06-2500 pg/mL
Catalog No. 50-000-81807 Supplier Meso Scale Discovery Supplier No. K151Z2S24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-2 Kit is a multiplex immunoassay that measures human interleukin-2 (IL-2) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay takes 3 5 hours to complete and has a detection range of 0 06-2500 pg/mL