Learn More
Meso Scale Discovery SPLEX HUM IL21 KitQP 25 PL
SDP
Catalog No. 5000079495
Encompass
The Meso Scale Discovery S-PLEX Human IL-21 Kit is an immunoassay that enables the simultaneous detection of IL-21 in human serum plasma and cell culture supernatant This multiplex assay utilizes electrochemiluminescent detection technology and is designed for use on Meso Scale Discovery instrumentation The kit includes a 96-well plate coated with capture antibodies specific for IL-21 detection antibodies conjugated to electrochemiluminescent labels calibrators and diluents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for IL-21 This product can aid in cytokine profiling and immune monitoring studies involving IL-21
Catalog No. 50-000-79495 Supplier Meso Scale Discovery Supplier No. K151ADNS24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-21 Kit is an immunoassay that enables the simultaneous detection of IL-21 in human serum plasma and cell culture supernatant This multiplex assay utilizes electrochemiluminescent detection technology and is designed for use on Meso Scale Discovery instrumentation The kit includes a 96-well plate coated with capture antibodies specific for IL-21 detection antibodies conjugated to electrochemiluminescent labels calibrators and diluents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for IL-21 This product can aid in cytokine profiling and immune monitoring studies involving IL-21