Learn More
Meso Scale Discovery SPLEX HUM IL22 KitQP 25 PL
SDP
Catalog No. 5000078656
Encompass
The Meso Scale Discovery S-PLEX Human IL-22 Kit is a multiplex immunoassay that allows simultaneous measurement of IL-22 in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrators and controls The assay takes approximately 3 5 hours to complete and has a detection range of 1 53 - 2500 pg/mL for IL-22
Catalog No. 50-000-78656 Supplier Meso Scale Discovery Supplier No. K151H3S24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-22 Kit is a multiplex immunoassay that allows simultaneous measurement of IL-22 in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer blocker solutions calibrators and controls The assay takes approximately 3 5 hours to complete and has a detection range of 1 53 - 2500 pg/mL for IL-22