Learn More
Meso Scale Discovery SPLEX HUM IL6 KitQP 25 PL
SDP
Catalog No. 5000081973
Encompass
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that enables the simultaneous detection of the human cytokine interleukin 6 (IL-6) using electrochemiluminescence (ECL) technology on the MSD platform This multiplex assay kit includes a plate coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG labels calibrators and an assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for IL-6 This product can be used for biomarker research and cytokine profiling in human serum plasma and cell culture supernatants
Catalog No. 50-000-81973 Supplier Meso Scale Discovery Supplier No. K151B3S24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that enables the simultaneous detection of the human cytokine interleukin 6 (IL-6) using electrochemiluminescence (ECL) technology on the MSD platform This multiplex assay kit includes a plate coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG labels calibrators and an assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for IL-6 This product can be used for biomarker research and cytokine profiling in human serum plasma and cell culture supernatants