Learn More
Meso Scale Discovery SPLEX HUM IL9 KitQP 25 PL
SDP
Catalog No. 5000080588
Encompass
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay for measuring IL-9 in human serum plasma and cell culture supernatant It uses electrochemiluminescence (ECL) detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators diluent wash buffer read buffer and plate seals The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument enabling precise IL-9 quantification with minimal hands-on time
Catalog No. 50-000-80588 Supplier Meso Scale Discovery Supplier No. K151R3S24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay for measuring IL-9 in human serum plasma and cell culture supernatant It uses electrochemiluminescence (ECL) detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators diluent wash buffer read buffer and plate seals The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument enabling precise IL-9 quantification with minimal hands-on time