Learn More
Meso Scale Discovery SPLEX HUM TNFB KitQP 25 PL
SDP
Catalog No. 5000080364
Encompass
The Meso Scale Discovery S-PLEX Human TNF- Kit is a multiplex immunoassay that measures TNF- levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is approximately 3 5 hours This is a useful research tool for measuring TNF- levels in various human sample types
Catalog No. 50-000-80364 Supplier Meso Scale Discovery Supplier No. K151ADPS24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human TNF- Kit is a multiplex immunoassay that measures TNF- levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is approximately 3 5 hours This is a useful research tool for measuring TNF- levels in various human sample types