Learn More
Meso Scale Discovery SPLEX Neurology PNL 1 human
SDP
Catalog No. 5000080925
Encompass
The Meso Scale Discovery S-Plex Neurology Panel 1 Human Kit is a multiplex immunoassay for measuring 8 neurology-related biomarkers in human serum plasma and CSF It includes assays for A 42 A 40 t-tau p-tau -synuclein NFL GFAP and YKL-40 Using electrochemiluminescent detection it offers high sensitivity and a broad dynamic range aiding in the analysis of Alzheimer s Parkinson s and other neurodegenerative disorders The kit contains a 96-well plate detection antibodies read buffer sample diluent calibrators and controls
Catalog No. 50-000-80925 Supplier Meso Scale Discovery Supplier No. K15639S4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-Plex Neurology Panel 1 Human Kit is a multiplex immunoassay for measuring 8 neurology-related biomarkers in human serum plasma and CSF It includes assays for A 42 A 40 t-tau p-tau -synuclein NFL GFAP and YKL-40 Using electrochemiluminescent detection it offers high sensitivity and a broad dynamic range aiding in the analysis of Alzheimer s Parkinson s and other neurodegenerative disorders The kit contains a 96-well plate detection antibodies read buffer sample diluent calibrators and controls