Learn More
Meso Scale Discovery SPLEX Neurology PNL 1 NHP Kit
SDP
Catalog No. 5000079301
Encompass
The Meso Scale Discovery S-Plex Neurology Panel 1 NHP Kit is a multiplex immunoassay for measuring alpha-synuclein amyloid beta 1-42 amyloid beta 1-40 tau and phosphorylated tau in non-human primate samples like CSF serum and plasma Using electrochemiluminescence detection it provides sensitive measurement of biomarkers linked to neurological diseases such as Alzheimer s and Parkinson s The kit includes a 96-well plate antibodies calibrators controls buffers blockers and seals for one plate Recommended samples are NHP CSF serum and plasma
Catalog No. 50-000-79301 Supplier Meso Scale Discovery Supplier No. K15640S2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-Plex Neurology Panel 1 NHP Kit is a multiplex immunoassay for measuring alpha-synuclein amyloid beta 1-42 amyloid beta 1-40 tau and phosphorylated tau in non-human primate samples like CSF serum and plasma Using electrochemiluminescence detection it provides sensitive measurement of biomarkers linked to neurological diseases such as Alzheimer s and Parkinson s The kit includes a 96-well plate antibodies calibrators controls buffers blockers and seals for one plate Recommended samples are NHP CSF serum and plasma