Learn More
Meso Scale Discovery SPLEX Neurology PNL 1 NHP
SDP
Catalog No. 5000080926
Encompass
The Meso Scale Discovery S-Plex Neurology Panel 1 (NHP) Kit is a multiplex immunoassay that measures biomarkers related to neurodegenerative diseases This kit allows for the simultaneous quantitative measurement of A 38 A 40 A 42 -synuclein p-tau181 and t-tau in human serum plasma and CSF The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer calibrator and controls The NHP kit enables efficient profiling of pathology-specific biomarkers associated with Alzheimer s disease Parkinson s disease and other neurodegenerative disorders
Catalog No. 50-000-80926 Supplier Meso Scale Discovery Supplier No. K15640S4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-Plex Neurology Panel 1 (NHP) Kit is a multiplex immunoassay that measures biomarkers related to neurodegenerative diseases This kit allows for the simultaneous quantitative measurement of A 38 A 40 A 42 -synuclein p-tau181 and t-tau in human serum plasma and CSF The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer calibrator and controls The NHP kit enables efficient profiling of pathology-specific biomarkers associated with Alzheimer s disease Parkinson s disease and other neurodegenerative disorders