Learn More
Meso Scale Discovery SPLEX NHP Neurofilament L
SDP
Catalog No. 5000081032
Encompass
The Simoa NF-Light Advantage Kit from Meso Scale Diagnostics is an ultrasensitive immunoassay for the quantitative measurement of neurofilament light chain (NfL) in human serum plasma and CSF The assay uses Meso Scale Diagnostics Simoa technology which enables detection of proteins at sub-femtomolar concentrations The assay has a lower limit of quantification of 0 04 pg/mL and a measuring range up to 1000 pg/mL The total assay time is 3 5 hours The kit includes a 96-well plate coated with anti-NfL capture antibody detection antibody calibrator and diluent The assay is intended for research use only and aids in the study of neurodegenerative diseases
Catalog No. 50-000-81032 Supplier Meso Scale Discovery Supplier No. K156AKGS4
Add to Cart
Add to Cart
Request a Quote
The Simoa NF-Light Advantage Kit from Meso Scale Diagnostics is an ultrasensitive immunoassay for the quantitative measurement of neurofilament light chain (NfL) in human serum plasma and CSF The assay uses Meso Scale Diagnostics Simoa technology which enables detection of proteins at sub-femtomolar concentrations The assay has a lower limit of quantification of 0 04 pg/mL and a measuring range up to 1000 pg/mL The total assay time is 3 5 hours The kit includes a 96-well plate coated with anti-NfL capture antibody detection antibody calibrator and diluent The assay is intended for research use only and aids in the study of neurodegenerative diseases