Learn More
Meso Scale Discovery SPLEX Proinflam PNL 1HMNKit
SDP
Catalog No. 5000082249
Encompass
The Meso Scale Discovery S-PLEX Proinflammatory Panel 1 (human) Kit is a multiplex immunoassay that simultaneously measures 10 human proinflammatory biomarkers The assay uses electrochemiluminescent detection technology and is run on Meso Scale Discovery instrumentation This kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The 10 analytes measured are IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- This assay allows high-throughput sensitive and reproducible quantification of multiple proinflammatory biomarkers from a small sample volume
Catalog No. 50-000-82249 Supplier Meso Scale Discovery Supplier No. K15396S1
The Meso Scale Discovery S-PLEX Proinflammatory Panel 1 (human) Kit is a multiplex immunoassay that simultaneously measures 10 human proinflammatory biomarkers The assay uses electrochemiluminescent detection technology and is run on Meso Scale Discovery instrumentation This kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The 10 analytes measured are IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- This assay allows high-throughput sensitive and reproducible quantification of multiple proinflammatory biomarkers from a small sample volume