Learn More
Meso Scale Discovery SPLEX SARSCoV2 N KitQP 25 PL
SDP
Catalog No. 5000080525
Encompass
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel for the simultaneous qualitative detection of nucleocapsid protein antigens for SARS-CoV-2 in human serum plasma and saliva This assay utilizes electrochemiluminescence (ECL) on the SECTOR Imager 2400 platform The kit includes a plate coated with capture antibodies detection antibodies conjugated to ruthenium and calibrators and controls needed to run the assay Results are obtained in under 3 hours This assay allows for high throughput SARS-CoV-2 antigen testing
Catalog No. 50-000-80525 Supplier Meso Scale Discovery Supplier No. K150ADHS24
Add to Cart
Add to Cart
Request a Quote
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel for the simultaneous qualitative detection of nucleocapsid protein antigens for SARS-CoV-2 in human serum plasma and saliva This assay utilizes electrochemiluminescence (ECL) on the SECTOR Imager 2400 platform The kit includes a plate coated with capture antibodies detection antibodies conjugated to ruthenium and calibrators and controls needed to run the assay Results are obtained in under 3 hours This assay allows for high throughput SARS-CoV-2 antigen testing