Learn More
Meso Scale Discovery SPLEX SARSCoV2 Spike
SDP
Supplier:  Meso Scale Discovery K150ADJS4
Encompass
The S-PLEX SARS-CoV-2 Spike Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that quantitatively measures antibodies specific to the SARS-CoV-2 spike protein This multiplex assay simultaneously measures IgG IgM and IgA antibodies and provides a semiquantitative result to aid in assessing recent or prior SARS-CoV-2 infection The kit includes a 96-well plate coated with recombinant SARS-CoV-2 spike protein as well as detection antibodies calibrators controls diluent and other necessary reagents Results are obtained in just a few hours using Meso Scale Diagnostics instrumentation and software
Catalog No. 50-000-78817
Add to Cart
Request a Quote