Learn More
Med Vet International Padded Precut Splint 3M Scotchcast Quick Step Double Sided Felt 2 X 10 inch Felt / Fiberglass White, 75210, 10/Case
Catalog No. 502829275
Encompass_Preferred
Water-repellant technology sheds water off the splint, helping keep the injured person's skin dry and reduces the risk of skin maceration Conforms easily – the seven-layer fiberglass interior offers conformability around the bends and curves of the human body Easy-open seams on both sides of the splint – the felt can be easily opened on either side of the splint for quick access to the fiberglass layers and trouble-free trimming Splinting roll delivery system helps prevent exposure to moisture, which causes product hardening, and as a result, waste 2 x 10 inches There are 10 padded precut splints in a box
Catalog No. 50-282-9275 Supplier Med Vet International Supplier No. 1193017CS
Water-repellant technology sheds water off the splint, helping keep the injured person's skin dry and reduces the risk of skin maceration Conforms easily – the seven-layer fiberglass interior offers conformability around the bends and curves of the human body Easy-open seams on both sides of the splint – the felt can be easily opened on either side of the splint for quick access to the fiberglass layers and trouble-free trimming Splinting roll delivery system helps prevent exposure to moisture, which causes product hardening, and as a result, waste 2 x 10 inches There are 10 padded precut splints in a box