Learn More
Med Vet International Padded Precut Splint 3M Scotchcast Quick Step Double Sided Felt 3 X 12 inch Felt / Fiberglass White, 75312, 10/Case
Catalog No. 502829276
Encompass_Preferred
Water-repellant so water sheds off splint, helping keep the skin dry; reduces the risk of skin maceration Seven-layer fiberglass of splint interior offers greater conformability around the bends and curves of the human body than one unified layer Easy-open seams on both sides of splint: The felt can be easily opened on either side of the splint for quick access to the fiberglass layers and trouble-free trimming Easy-to-use, easy-to-reseal packaging helps prevent exposure to moisture, which causes product hardening, and as a result, waste 3 x 12 inches
Catalog No. 50-282-9276 Supplier Med Vet International Supplier No. 1193018CS
Water-repellant so water sheds off splint, helping keep the skin dry; reduces the risk of skin maceration Seven-layer fiberglass of splint interior offers greater conformability around the bends and curves of the human body than one unified layer Easy-open seams on both sides of splint: The felt can be easily opened on either side of the splint for quick access to the fiberglass layers and trouble-free trimming Easy-to-use, easy-to-reseal packaging helps prevent exposure to moisture, which causes product hardening, and as a result, waste 3 x 12 inches