Learn More
Med Vet International Padded Precut Splint 3M Scotchcast Quick Step Double Sided Felt 3 X 35 inch Felt / Fiberglass White, 75335, 10/Case
Catalog No. 502829277
Encompass_Preferred
Water-repellant technology sheds water off the splint, helping keep the injured person's skin dry and reduces the risk of skin maceration Conforms easily – the seven-layer fiberglass interior offers conformability around the bends and curves of the human body Easy-open seams on both sides of the splint – the felt can be easily opened on either side of the splint for quick access to the fiberglass layers and trouble-free trimming Splinting roll delivery system helps prevent exposure to moisture, which causes product hardening, and as a result, waste 3 x 35 inches There are 10 padded precut splints in a carton
Catalog No. 50-282-9277 Supplier Med Vet International Supplier No. 1193019CS
Water-repellant technology sheds water off the splint, helping keep the injured person's skin dry and reduces the risk of skin maceration Conforms easily – the seven-layer fiberglass interior offers conformability around the bends and curves of the human body Easy-open seams on both sides of the splint – the felt can be easily opened on either side of the splint for quick access to the fiberglass layers and trouble-free trimming Splinting roll delivery system helps prevent exposure to moisture, which causes product hardening, and as a result, waste 3 x 35 inches There are 10 padded precut splints in a carton