Learn More
Med Vet International Padded Splint Roll 3M Scotchcast Quick Step Double Sided Felt 2 inch X 15 Foot Felt / Fiberglass White, 74002Q, 2/Case
Catalog No. 502830636
Encompass_Preferred
Felt on both sides of splint helps eliminate the risk of wrong-side application while providing padding and protection for injury Water-repellant technology sheds water off splint, helping keep skin dry and reduce the risk of skin maceration Superior conformability – the seven-layer fiberglass of splint interior offers greater conformability than one unified layer Easy-open seams on both sides of splint: The felt can be easily opened on either side of the splint for quick access to the fiberglass layers and trouble-free trimming 2 inches x 15 feet
Catalog No. 50-283-0636 Supplier Med Vet International Supplier No. 983389CS
Felt on both sides of splint helps eliminate the risk of wrong-side application while providing padding and protection for injury Water-repellant technology sheds water off splint, helping keep skin dry and reduce the risk of skin maceration Superior conformability – the seven-layer fiberglass of splint interior offers greater conformability than one unified layer Easy-open seams on both sides of splint: The felt can be easily opened on either side of the splint for quick access to the fiberglass layers and trouble-free trimming 2 inches x 15 feet