Learn More
Med Vet International Padded Splint Roll 3M Scotchcast Quick Step Double Sided Felt 4 inch X 15 Foot Felt / Fiberglass White, 74004Q, 2/Case
Catalog No. 502830638
Encompass_Preferred
The white 3M Scotchcast splint has felt on both sides to avoid wrong-side applications It has seams on both sides for quick access It comes in easy-to-use, resealable packaging to minimize the risk of moisture exposure that can harden the cast splint The fiberglass splint conforms around body contours to provide effective protection It sheds water to protect them skin from becoming wrinkly and soggy A case contains two 4 inch x 15 feet casts
Catalog No. 50-283-0638 Supplier Med Vet International Supplier No. 983391CS
The white 3M Scotchcast splint has felt on both sides to avoid wrong-side applications It has seams on both sides for quick access It comes in easy-to-use, resealable packaging to minimize the risk of moisture exposure that can harden the cast splint The fiberglass splint conforms around body contours to provide effective protection It sheds water to protect them skin from becoming wrinkly and soggy A case contains two 4 inch x 15 feet casts