Learn More
Med Vet International Miltex Vantage Splinter Liberator, 5in.
Catalog No. 502829575
Encompass_Preferred
The Miltex Vantage V96-400 Splinter Liberator is made of mid-grade stainless steel and has a length of 5in. It has a reversible handle with needle sharp tips.
Catalog No. 50-282-9575 Supplier Med Vet International Supplier No. MILV96400
The Miltex Vantage V96-400 Splinter Liberator is made of mid-grade stainless steel and has a length of 5in. It has a reversible handle with needle sharp tips.