Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design Splinter Tweezer Forceps, Fine Pointed, Serrated Jaws

Catalog No. 22079760 Shop All Surgical Design Products
Change view
Click to view available options
Length (Metric):
8.89 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
22-079-760 8.89 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-079-760 Supplier Surgical Design Supplier No. 256
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Surgical Design™ Splinter Forceps are multi-purpose forceps used in first aid, research experiments, general surgery and more. The advantageous fine tips are primarily used to extract small, foreign objects on the surface of the skin. It is also used for handling bandages, sutures, cotton/gauze, and more.

  • Used in variety of applications
  • Easy and effortless grasping of fine tissue
  • Lightweight and portable
  • High polish finish
  • Available in various sizes and curvatures
  • Specifications

    Product Type Forceps, Splinter, Fine-point
    Finish Polished
    For Use With (Application) Multipurpose
    Grade Economy
    Quantity 12
    Tip Style Straight, Serrated
    Length (English) 3.5 in.
    Length (Metric) 8.89 cm
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.