Learn More
Med Vet International Plastic Splints Medium 1.5"X 12", J0119B
Catalog No. 502831850
Encompass_Preferred
Made of strong, light-weight plastic and padded with non-irritating foam. Can be contoured at distal end with addition of moderate heat; modified cutout for carpal pad. Radiolucent- will not interfere with X-ray while in place. Familiar green color.
Catalog No. 50-283-1850 Supplier Med Vet International Supplier No. JOR119B
Made of strong, light-weight plastic and padded with non-irritating foam. Can be contoured at distal end with addition of moderate heat; modified cutout for carpal pad. Radiolucent- will not interfere with X-ray while in place. Familiar green color.