Learn More
Roboz Surgical Roboz Surgical Instrument Co. Splitable Introducer, 17 Gauge, Red, 10/Pk.
SDP
Catalog No. 5000574431
Encompass
This splitable introducer cannula is designed for precise vascular access applications. It features a 17-gauge diameter and is color-coded red for easy identification in clinical or laboratory settings.
  • Features a 1.5 mm outside diameter
  • Designed with a 1.34 mm inside diameter
  • Supplied in a pack of 10
Catalog No. 50-005-74431 Supplier Roboz Surgical Supplier No. IN517
This splitable introducer cannula is designed for precise vascular access applications. It features a 17-gauge diameter and is color-coded red for easy identification in clinical or laboratory settings.
  • Features a 1.5 mm outside diameter
  • Designed with a 1.34 mm inside diameter
  • Supplied in a pack of 10