Learn More
Cayman Chemical Splitomicin, Purity: ≥98%, CAS: 5690-03-9, 10 mg
SDP
Catalog No. 502952839
Encompass_Preferred
A small molecule inhibitor of Sir2p HDAC activity, displaying higher activity in vivo (minimal inhibitory concentration = 0.49 µM) than in vitro (IC50 = 60 µM); has diverse effects on mammalian cells
Catalog No. 50-295-2839 Supplier Cayman Chemical Supplier No. 1316810 MG
A small molecule inhibitor of Sir2p HDAC activity, displaying higher activity in vivo (minimal inhibitory concentration = 0.49 µM) than in vitro (IC50 = 60 µM); has diverse effects on mammalian cells