Learn More
AdipoGen Splitomicin (1 mg)
SDP
Supplier:  AdipoGen AGCR10088M001
Encompass_Preferred
Chemical. CAS: 5690-03-9. Formula: C13H10O2. MW: 198.2. Potent cell permeable and selective inhibitor of yeast NAD+-dependent histone deacetylase (HDAC) Sir2p. Displays higher activity in vivo than in vitro. Sensitizes mammalian cells to a variety of DNA-damaging agents by abrogating Sir2p activity on p53. Acts by either altering or blocking access to the acetylated histone binding pocket. Shown to have diverse effects also in mammalian cells.
Catalog No. 50-205-3828