Learn More
Medchemexpress LLC Splitomicin | 5690-03-9 | MFCD08705254 | 98.2% | 198.22 g/mol | C13H10O2 | 100 MG
SDP
Catalog No. 5000213652
Encompass_Preferred
Splitomicin is a selective Sir2p (sirtuin) inhibitor used as a research reagent to study NAD+-dependent histone deacetylase activity and related epigenetic pathways. Supplied as a purified small-molecule, it is intended for in vitro biochemical assays and pathway investigations.
  • Selective Sir2p inhibitor to probe NAD+-dependent HDAC activity.
  • High reported purity (≈98%) suitable for biochemical assays.
  • Molecular weight 198.22 g/mol with formula C13H10O2.
  • Available in small pack sizes appropriate for research use, including 100 mg.
  • Commonly used in sirtuin pathway and epigenetics studies.
Catalog No. 50-002-13652 Supplier Medchemexpress LLC Supplier No. HY100585100MG
Splitomicin is a selective Sir2p (sirtuin) inhibitor used as a research reagent to study NAD+-dependent histone deacetylase activity and related epigenetic pathways. Supplied as a purified small-molecule, it is intended for in vitro biochemical assays and pathway investigations.
  • Selective Sir2p inhibitor to probe NAD+-dependent HDAC activity.
  • High reported purity (≈98%) suitable for biochemical assays.
  • Molecular weight 198.22 g/mol with formula C13H10O2.
  • Available in small pack sizes appropriate for research use, including 100 mg.
  • Commonly used in sirtuin pathway and epigenetics studies.