Learn More
Medchemexpress LLC HY-100585 5mg Medchemexpress, Splitomicin CAS:5690-03-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1005855MG
Encompass_Preferred
Medchemexpress, HY-100585 5mg Splitomicin CAS:5690-03-9 Splitomicin (Splitomycin) is a selective Sir2p inhibitor. Splitomicin inhibits NAD+-dependent HDAC activity of Sir2 protein. Splitomicin induces dose-dependent inhibition of HDAC in the yeast extract with an IC50 of 60 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7750