Learn More
Perkin Elmer US LLC Split/Splitless Injector Starter Kit for Manual Gas Control
Catalog No. 502104017
Encompass_Preferred
Includes the following pieces: (5) Ultra Inert Splitless Liners with Wool 4 mm I.D. (09200696) (5) Ultra Deactivated Split/Splitless Liners with Glass Wool, 4 mm I.D. (09200624) (10) Silicone O-Rings (09200714) (50) Green Injection Port Septa (N6621028) (10) 0.4 mm (09200685), (10) 0.5 mm (09200686) and (10) 0.8 mm Graphite/Vespel Ferrules (09200687) (2) Gold Seal Washers (N6551043) (1) Gold Seal Nut (N6552080) (1) Capillary Injector Nut (N6552084) (1) Ceramic Column Cutter (N9301376)
Catalog No. 50-210-4017 Supplier Perkin Elmer US LLC Supplier No. N6550101
Includes the following pieces: (5) Ultra Inert Splitless Liners with Wool 4 mm I.D. (09200696) (5) Ultra Deactivated Split/Splitless Liners with Glass Wool, 4 mm I.D. (09200624) (10) Silicone O-Rings (09200714) (50) Green Injection Port Septa (N6621028) (10) 0.4 mm (09200685), (10) 0.5 mm (09200686) and (10) 0.8 mm Graphite/Vespel Ferrules (09200687) (2) Gold Seal Washers (N6551043) (1) Gold Seal Nut (N6552080) (1) Capillary Injector Nut (N6552084) (1) Ceramic Column Cutter (N9301376)