Learn More
IDEX Health & Science Upchurch SPLITTER ASSEMBLY MICRO 1/4-28
Catalog No. NC1919708
Encompass_Preferred
Idex Micro-Metering and Micro-Splitter Valves: Needle Valves, Flangeless Nuts and Ferrules; P-200, P-235 Fitings, Pressure: 800, PEEK, For Use With: Chromatography Columns, Swept Volume: 4.1 uL Fully Open
Catalog No. NC1919708 Supplier IDEX Health & Science Upchurch Supplier No. P450
Idex Micro-Metering and Micro-Splitter Valves: Needle Valves, Flangeless Nuts and Ferrules; P-200, P-235 Fitings, Pressure: 800, PEEK, For Use With: Chromatography Columns, Swept Volume: 4.1 uL Fully Open