Learn More
Waters Corp Splitter Assembly, PEEK, for UPC2/MS Systems
Catalog No. 502391184 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a replacement splitter assembly designed for use with UPC2/MS systems. It is constructed from PEEK material.
  • Made from PEEK material.
  • Designed for UPC2/MS systems.
  • Serves as a replacement part.
Catalog No. 50-239-1184 Supplier Waters Corp Supplier No. 205001683
This product is a replacement splitter assembly designed for use with UPC2/MS systems. It is constructed from PEEK material.
  • Made from PEEK material.
  • Designed for UPC2/MS systems.
  • Serves as a replacement part.