Learn More
Ace Glass, Inc. Splitter, distillation, reflux, 29/32 top & bottom joints, 0-10mmside valve, 1/2-in ground outlet
SDP
Catalog No. 50972758
Encompass_Preferred
Splitter, distillation, reflux, 29/32 top & bottom joints, 0-10mmside valve, 1/2-in ground outlet
Catalog No. 50-972-758 Supplier Ace Glass, Inc. Supplier No. 608812
Splitter, distillation, reflux, 29/32 top & bottom joints, 0-10mmside valve, 1/2-in ground outlet