Learn More
Neta Scientific Agilent Split vent trap PM kit,single cartridge
SDP
Catalog No. 50924421
Encompass_Preferred
This item is manufactured by Agilent
Agilent Split vent trap preventative maintenance kit for split/splitless, volatiles, and programmable temperature vaporizing (PTV) septumless inlet split vent line. Includes cartridge, 2 O-rings
- AA
Catalog No. 50-924421 Supplier Neta Scientific Supplier No. 51886495
This item is manufactured by Agilent
Agilent Split vent trap preventative maintenance kit for split/splitless, volatiles, and programmable temperature vaporizing (PTV) septumless inlet split vent line. Includes cartridge, 2 O-rings
- AA