Learn More
Cayman Chemical Spltomcn 5mg
SDP
Supplier:  Cayman Chemical 131685 MG
Encompass_Preferred
A small molecule inhibitor of Sir2p HDAC activity, displaying higher activity in vivo (minimal inhibitory concentration = 0.49 µM) than in vitro (IC50 = 60 µM); has diverse effects on mammalian cells
Catalog No. 50-295-1748