Learn More
Med Vet International Two-in-One Combination Pill Splitter and Crusher, 5 Pairs, J1322
Catalog No. 5000495656
Encompass_Preferred
Two-in-One Combination Pill-Splitter and Crusher Crush Cups. Two cups are used by simply placing the tablet into the bottom of the first cup. It has a raised pointed grid pattern inside on the bottom. A second cup has a raised pointed grid pattern on the outside of the bottom of the cup. Placing the second cup in the first cup and rotating cups in opposite direction will quickly grind the tablet into a uniform powder. Dimensions: 2"(50mm) across the top x 1"(25mm) across the bottom with 1 3/4"(44mm) tall. Material is thick, white, opaque plastic. 5 sets of 2 cups/bag
Catalog No. 50-004-95656 Supplier Med Vet International Supplier No. JOR1322
Two-in-One Combination Pill-Splitter and Crusher Crush Cups. Two cups are used by simply placing the tablet into the bottom of the first cup. It has a raised pointed grid pattern inside on the bottom. A second cup has a raised pointed grid pattern on the outside of the bottom of the cup. Placing the second cup in the first cup and rotating cups in opposite direction will quickly grind the tablet into a uniform powder. Dimensions: 2"(50mm) across the top x 1"(25mm) across the bottom with 1 3/4"(44mm) tall. Material is thick, white, opaque plastic. 5 sets of 2 cups/bag