Learn More
Med Vet International Two-in-One Combination Pill Splitter and Crusher, J1322B, 20 Sets
Catalog No. 5000495739
Encompass_Preferred
Two-in-One Combination Pill-Splitter and Crusher Crush Cups. Two cups are used by simply placing the tablet into the bottom of the first cup. It has a raised pointed grid pattern inside on the bottom. A second cup has a raised pointed grid pattern on the outside of the bottom of the cup. Placing the second cup in the first cup and rotating cups in opposite direction will quickly grind the tablet into a uni- form powder. Dimensions: 2"(50mm) across the top x 1"(25mm) across the bottom with 1 3/4"(44mm) tall. Material is thick, white, opaque plastic. Pill Splitter Cup. Two cup system where bottom of the first cup has a triangular space for holding the tablet. The second cup has sharp midline edging. Place the tablet in the bottom of the cup and press the second cup on top tablet splitting it. Dimensions: 2"(50mm) across the top x 1"(25mm) across the bottom with 1 3?4"(44mm) tall. 20 sets of 2 cups/bag
Catalog No. 50-004-95739 Supplier Med Vet International Supplier No. JOR1322B
Two-in-One Combination Pill-Splitter and Crusher Crush Cups. Two cups are used by simply placing the tablet into the bottom of the first cup. It has a raised pointed grid pattern inside on the bottom. A second cup has a raised pointed grid pattern on the outside of the bottom of the cup. Placing the second cup in the first cup and rotating cups in opposite direction will quickly grind the tablet into a uni- form powder. Dimensions: 2"(50mm) across the top x 1"(25mm) across the bottom with 1 3/4"(44mm) tall. Material is thick, white, opaque plastic. Pill Splitter Cup. Two cup system where bottom of the first cup has a triangular space for holding the tablet. The second cup has sharp midline edging. Place the tablet in the bottom of the cup and press the second cup on top tablet splitting it. Dimensions: 2"(50mm) across the top x 1"(25mm) across the bottom with 1 3?4"(44mm) tall. 20 sets of 2 cups/bag