Learn More
Cayman Chemical SPM D-series MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502964991
Encompass_Preferred
Contains several D-series resolvins and their precursor, DHA; designed for direct use in LC-MS applications
Catalog No. 50-296-4991 Supplier Cayman Chemical Supplier No. 187021 EA
Contains several D-series resolvins and their precursor, DHA; designed for direct use in LC-MS applications