Learn More
Cayman Chemical SPM E-series MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502984443
Encompass_Preferred
Contains two E-series resolvins and their precursor, EPA; designed for direct use in LC-MS applications
Catalog No. 50-298-4443 Supplier Cayman Chemical Supplier No. 194171 EA
Contains two E-series resolvins and their precursor, EPA; designed for direct use in LC-MS applications