Learn More
Trajan Scientific CTC SPME Arrow Liner
Catalog No. NC1917200
Encompass
CTC SPME Arrow Liner ID 1.7mm, for SSL Injector of Thermo GC Trace1300, Pack - 3 For any SPME Arrow, PAL, PAL3
Catalog No. NC1917200 Supplier Trajan Scientific Supplier No. ARRLIN17TRACE13003
CTC SPME Arrow Liner ID 1.7mm, for SSL Injector of Thermo GC Trace1300, Pack - 3 For any SPME Arrow, PAL, PAL3