Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carbowax-Polyethylene Glycol (PEG) Coating, df 60 μm
Catalog No. 111008235 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-100-8235 Autosampler
11-100-8248 Manual Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-8235 Supplier MilliporeSigma™ Supelco™ Supplier No. 57354U
Add to Cart
Add to Cart
Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Plain purple hub
Needle Gauge 23 ga
Quantity 3/Pk.