Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm
Catalog No. 111020749 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
Needle Gauge:
23 ga
24 ga
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Needle Gauge
11-102-0749 Autosampler 23 ga
11-102-0753 Autosampler 24 ga
11-100-8485 Manual Holder 23 ga
11-100-8581 Manual Holder 24 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-0749 Supplier MilliporeSigma™ Supelco™ Supplier No. 57343U
Add to Cart
Add to Cart
For Gases and Low Molecular Weight Compounds (MW 30-225)
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 23 ga
Quantity 3/Pk.