Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm
Catalog No. 111007790 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
Material:
Metal Alloy Fiber
StableFlex Fiber
Needle Gauge:
23 ga
24 ga
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material Needle Gauge
11-100-7790 Autosampler Metal Alloy Fiber 23 ga
11-102-0743 Autosampler StableFlex Fiber 23 ga
11-100-8573 Autosampler StableFlex Fiber 24 ga
11-100-8583 Manual Holder StableFlex Fiber 24 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-100-7790 Supplier MilliporeSigma™ Supelco™ Supplier No. 57906U
Add to Cart
Add to Cart
For Gases and Low Molecular Weight Compounds (MW 30-225)
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Metal Alloy Fiber
Needle Gauge 23 ga
Quantity 1/Pk.