Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Divinylbenzene/Carboxen™/Polydimethylsiloxane (DVB/CAR/PDMS), df 50/30 μm
Catalog No. 111020756 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Needle Gauge:
23 ga
23 ga, L: 1 cm
24 ga
For Use With (Equipment):
Autosampler
Manual Holder
Description:
Notched Gray Hub
Plain Gray Hub
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge For Use With (Equipment) Description
11-102-0756 24 ga Manual Holder Notched Gray Hub
11-102-0745 23 ga Autosampler -
11-102-0754 24 ga Autosampler -
11-102-0746 23 ga Manual Holder -
11-102-0755 24 ga Manual Holder Plain Gray Hub
11-100-7791 23 ga, L: 1 cm Autosampler -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 11-102-0756 Supplier MilliporeSigma™ Supelco™ Supplier No. 57348U
Add to Cart
Add to Cart
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS) is manufactured with a flexible metal alloy used in the needle, plunger, and fiber core.
The new metal alloy design includes a thicker, flexible plunger that is much less likely to kink or break, and helps to reinforce the needle especially when used in an auto-sampler with a sample agitator. Since the needle is more flexible and has a thinner wall than the standard stainless steel needle, a bevel has been placed on the needle to help it pierce septa materials more easily.
TRUSTED_SUSTAINABILITY
Specifications
Description Notched Gray Hub
For Use With (Application) SPME
For Use With (Equipment) Manual Holder
Type SPME Fibers and Accessories
Material StableFlex Fiber
Needle Gauge 24 ga
Quantity 3/Pk.
Carboxen and StableFlex are registered trademarks of Sigma-Aldrich Co. LLC