Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm, StableFlex™
Catalog No. 111020744 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
Material:
Fused Silica Fiber
Plain pink hub
Polydimethylsiloxane/Divinylbenzene
Needle Gauge:
23 ga
24 ga
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material Needle Gauge
11-102-0744 Autosampler Polydimethylsiloxane/Divinylbenzene 23 ga
11-100-8572 Autosampler Plain pink hub 24 ga
11-100-8486 Manual Holder Fused Silica Fiber 23 ga
11-100-8582 Manual Holder Plain pink hub 24 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-0744 Supplier MilliporeSigma™ Supelco™ Supplier No. 57293U
Add to Cart
Add to Cart
Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Polydimethylsiloxane/Divinylbenzene
Needle Gauge 23 ga
Quantity 3/Pk.
StableFlex is a trademark of Sigma-Aldrich Co. LLC