Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Polydimethylsiloxane (PDMS), df 100 μm
Catalog No. 111020750 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
Material:
Fused Silica Fiber
Metal Alloy Fiber
Needle Gauge:
23 ga
24 ga
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material Needle Gauge
11-102-0750 Autosampler Fused Silica Fiber 23 ga
11-100-7705 Autosampler Metal Alloy Fiber 23 ga
11-100-8570 Autosampler Fused Silica Fiber 24 ga
11-100-8484 Manual Holder Fused Silica Fiber 23 ga
11-101-6270 Manual Holder Fused Silica Fiber 24 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-102-0750 Supplier MilliporeSigma™ Supelco™ Supplier No. 57341U
Add to Cart
Add to Cart
Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 23 ga
Quantity 3/Pk.