Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Polydimethylsiloxane (PDMS), df 7 μm
Catalog No. 111008233 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
Needle Gauge:
23 ga
24 ga
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Needle Gauge
11-100-8233 Autosampler 23 ga
11-100-8489 Autosampler 24 ga
11-100-8501 Manual Holder 24 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-8233 Supplier MilliporeSigma™ Supelco™ Supplier No. 57291U
Add to Cart
Add to Cart
Solid phase microextraction technique can be used for GC/MS analysis. It helps in simplifying and speeds up the sample preparation. It also reduces solvent requirement for the extraction.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 23 ga
Quantity 3/Pk.