Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assortment Kit 2
Catalog No. 111008602 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
Needle Gauge:
23 ga
24 ga
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Needle Gauge
11-100-8602 Manual Holder 24 ga
11-100-8237 Autosampler 23 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-8602 Supplier MilliporeSigma™ Supelco™ Supplier No. 57320U
Add to Cart
Add to Cart
Kit 2 - For Volatile or Polar Organics in Water - contains one of each: 75 μm Carboxen/polydimethylsiloxane coating, 65 μm polydimethylsiloxane/divinylbenzene coating and 85 μm Polyacrylate coating
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Manual Holder
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 24 ga