Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assortment Kit 2
Catalog No. 111008574 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Catalog No. For Use With (Equipment)
11-100-8574 Autosampler
1 options
Catalog No. 11-100-8574 Supplier MilliporeSigma™ Supelco™ Supplier No. 57321U
Add to Cart
Add to Cart
Kit 2 - For Volatile or Polar Organics in Water - contains one of each: 75 μm Carboxen/polydimethylsiloxane coating, 65 μm polydimethylsiloxane/divinylbenzene coating and 85 μm Polyacrylate coating
TRUSTED_SUSTAINABILITY
Specifications
Description For analyte group volatile and polar organics aq. soln.
For Use With (Application) Food and Beverages
For Use With (Equipment) Autosampler
Type SPE Accessory
Material Fused silica fiber
Needle Gauge 24 ga