Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assortment Kit 4
Catalog No. 111008575 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Manual Holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-100-8575 Autosampler
11-100-8585 Manual Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-8575 Supplier MilliporeSigma™ Supelco™ Supplier No. 57325U
Add to Cart
Add to Cart
Kit 4 - For Flavors and Odors - contains one of each: 100 μm polydimethylsiloxane coating, 65 μm polydimethylsiloxane/divinylbenzene coating and 75 μm Carboxen/polydimethylsiloxane coating
TRUSTED_SUSTAINABILITY
Specifications
Description For analyte group flavors and odors
For Use With (Application) Food and Beverages
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Fused silica fiber
Needle Gauge 24 ga