Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assortment Kit 4
Catalog No. 111008238 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Catalog No. For Use With (Equipment)
11-100-8238 Autosampler
1 options
Catalog No. 11-100-8238 Supplier MilliporeSigma™ Supelco™ Supplier No. 57287U
Add to Cart
Add to Cart
Kit 4 - For Flavors and Odors - contains one of each: 100 μm polydimethylsiloxane coating, 65 μm polydimethylsiloxane/divinylbenzene coating and 75 μm Carboxen/polydimethylsiloxane coating
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 23 ga