Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assortment Kit 5
Catalog No. 111007317 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Catalog No. For Use With (Equipment)
11-100-7317 Autosampler
1 options
Catalog No. 11-100-7317 Supplier MilliporeSigma™ Supelco™ Supplier No. 57362U
Add to Cart
Add to Cart
SPME Fiber Assortment Kit 5 contains one fiber of each: 65 μm PDMS/DVB coating, 50/30 μm DVB/Carboxen/PDMS coating, 85 μm Carboxen/PDMS coating and 100 μm PDMS coating
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material Fused Silica Fiber
Needle Gauge 23 ga