Learn More
MilliporeSigma™ Supelco™ SPME Fiber Holder
Catalog No. 111020752 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
CTC CombiPAL, Gerstel MPS2 and Thermo TriPlus Autosamplers
Manual Sampling
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-102-0752 CTC CombiPAL, Gerstel MPS2 and Thermo TriPlus Autosamplers
11-101-6228 Manual Sampling
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0752 Supplier MilliporeSigma™ Supelco™ Supplier No. 57347U
Add to Cart
Add to Cart
Use this holder with SPME fiber assemblies that are designed for automated sampling.
An adjustable depth guide positions the fiber for sampling and for correct placement in the heated zone of the GC injection port. The fiber can be locked in the exposed position.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Food and Beverages
For Use With (Equipment) CTC CombiPAL, Gerstel MPS2 and Thermo TriPlus Autosamplers
Type SPME Fibers and Accessories